Human MG (from Urine (post-menopausal women))
: Biorbyt
- Ursprung:Urine (post-menopausal women)
- Spezies:Human
- Lagerbedingungen:Lyophilized Menopausal Gonadotropin Human although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution MG should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Biologische Aktivität:100 IU/mg of FSH and 100 IU/mg LH.
- Protein/Peptid-Name:MG
- Purity:Greater than 95.0% as determined by SDS-PAGE.
- Solubility:It is recommended to reconstitute the lyophilized MG in sterile H2O not less than 100 ug/ml, which can then be further diluted to other aqueous solutions.
- Zusammensetzung:The Human MG was lyophilized from concentrated (1 mg/ml) solution with no additives.
- Validierte Anwendungen:SDS-PAGE
Frequently Bought Together