ContactUsLinkComponent
Mouse recombinant IL9
Lieferant:  Biorbyt
Zertifikate
Mouse recombinant IL9
Lieferant:  Biorbyt
Spezifikationen
  • Protein/Peptid-Typ:
    Rekombinant
  • Spezies:
    Mouse
  • Lagerbedingungen:
    Store at −20 °C.
  • Biologische Aktivität:
    The ED50 as determined by the dose-dependent proliferation of human MO7e cells is < 0.5 ng/ml, corresponding to a specific activity of > 2 x 106 units/mg.
  • Proteinsynonym:
    p40 cytokine
  • UniProtKB:
    P15247
  • Protein/Peptid-Name:
    IL9
  • Purity:
    >98%
  • Solubility:
    It is recommended to reconstitute the lyophilized Interleukin in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    MQRCSTTWGI RDTNYLIENL KDDPPSKCSC SGNVTSCLCL SVPTDDCTTP CYREGLLQLT NATQKSRLLP VFHRVKRIVE VLKNITCPSF SCEKPCNQTM AGNTMSFLKS LLGTFQKTEM QRQKSRP
  • Zusammensetzung:
    Lyophilized from concentrated (1mg/ml) solution containing no additives.
  • Validierte Anwendungen:
    SDS-PAGE
Spezifikationen
Zugehörige Informationen
Frequently Bought Together