Mouse recombinant IL15
Lieferant: Biorbyt
Zertifikate
Über diesen Artikel
Spezifikationen
- Protein/Peptid-Typ:Rekombinant
- Spezies:Mouse
- Lagerbedingungen:Store at −20 °C.
- Biologische Aktivität:The ED50 was determined by the stimulation of the proliferation of Mouse CTLL-2 cells is < 5.0 ng/ml, corresponding to a specific activity of > 2 x 105 units/mg.
- Proteinsynonym:IL-T
- UniProtKB:P48346
- Protein/Peptid-Name:IL15
- Purity:>98%
- Solubility:It is recommended to reconstitute the lyophilized Interleukin-15 in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:MNWIDVRYDL EKIESLIQSI HIDTTLYTDS DFHPSCKVTA MNCFLLELQV ILHEYSNMTL NETVRNVLYL ANSTLSSNKN VAESGCKECE ELEEKTFTEF LQSFIRIVQM FINTS
- Zusammensetzung:The protein was lyophilized from concentrated (1mg/ml) solution with 10mM NaP pH 8.0.
- Validierte Anwendungen:SDS-PAGE