Protein and serum free medium, PF medium, Corning®
Artikelnr. MDTC40-200-CV
Supplier: Corning
CAS Number:
Specifications
- Medienformat:Flüssigkeit
- VE:6
- Bezeichnung:PF medium, without L-glutamine, 6×500 ml
Specifications
About this item
PF medium is optimised for the cultivation of Chinese Hamster Ovary (CHO) cells, along with many derivatives of this parent line PF medium is a proprietary serum-free and protein-free growth medium that does not contain any hormones or growth factors. It medium supports superior growth and viability over long term passages in both adherent and suspension culture, and is formulated without L-glutamine.