AuftragsverfolgungKontakt
MSD conversion fitting | Avantor
MSD conversion fitting
MSD conversion fitting, Messing, vernickelt
Artikelnr. REST21314
Anbieter:   Restek
MSD conversion fitting
Artikelnr. REST21314
Anbieter:  Restek
MSD conversion fitting
MSD conversion fitting
MSD conversion fitting, Messing, vernickelt
Preis
74,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
This conversion fitting is a flat, soft aluminum sealing ring deforms and butt-seals against the MSD interface. A standard Vespel® ferrule seals the column and 1,59 mm stainless steel nut.
  • Fitting is constructed of nickel-plated brass for longevity and softness
  • Use any standard Vespel® or Vespel®/graphite 1,59 mm ferrule
  • Improved design reduces chance of leaks
Lieferumfang: This fitting includes a 1,59 mm stainless steel nut and two replacement sealing rings. Ferrules have to be ordered separately.
Spezifikationen
  • Bezeichnung:
    MSD conversion fitting
  • Material:
    Messing, vernickelt
Accessories
Produktinformationen
  • Artikelnr.
    Bezeichnung
    Verfügbarkeit
    Preis
  • REST21313
    Replacement ring seal for MSD conversion fitting
    Einloggen für Verfügbarkeit
    1 * 2 ST - 24,00 €