Mega 108 Storage Cabinet for HPLC Columns
Artikelnr. TRNT50001
Supplier: TRIPP
Specifications
- Bezeichnung:HPLC storage cabinet, with clear lids
- B×T×H:381×406×406 mm
- VE:1
- Gewicht:13 kg
Specifications
About this item
HPLC storage cabinet with constructed with white PVC and acrylic doors.
- Mega organiser cabinet with seven removable, slide-out trays
- Cabinet has a total of 108 adjustable compartments
- Removable dividers fit most analytical HPLC columns
- Each tray is covered with an acrylic lid that is completely removable
Ordering information: Doors with different colours are available on request.