Order Entry
Austria
ContactUsLinkComponent
Anti-MSTN Rabbit Polyclonal Antibody
Anti-MSTN Rabbit Polyclonal Antibody
Catalog # BOSSBS-1288R
Supplier:  Bioss
undefined
Anti-MSTN Rabbit Polyclonal Antibody
Catalog # BOSSBS-1288R
Supplier:  Bioss

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Myostatin
  • Antigen-Symbol:
    MSTN
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • ImmunFluoreszenz:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Kreuzadsorption:
    No
  • Form:
    Liquid
  • Gen-ID:
    2660
  • Antigen-Synonyme:
    GDF8|CMPT|MSLHP
  • Lagerungspuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Lagertemperatur:
    Store at –20 °C for 12 months
  • Konzentration:
    1 ug/ul
  • Transporttemperatur:
    4 °C
  • Immunogen:
    300-350/375
  • Reinigung:
    Purified by Protein A
  • Größe:
    100 μL
  • VE:
    100 µl

Specifications

About this item

Type: Primary
Antigen: MSTN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat