Micropacked GC columns
Lieferant: Restek
Zertifikate
Über diesen Artikel
These micropacked columns are highly efficient and provide good sample capacity. The unsurpassed inertness of SilcoSmooth tubing is based on Siltek deactivation, which allows the column to be flexed and coiled without any fear of chipping or cracking the inert surface.
- Efficient, inert, and flexible
- Easy to install due to multiple internal diameters
- Standard coils fit all instruments
- No special instrument configurations required
- Micropacked columns operate at flows exceeding 10 ml/min for trouble-free operation
Spezifikationen
- Catalog No:
- REST19012
- REST19013
- REST19014
- REST19015
- REST19009
- REST19010
- REST19011
- REST19002-293
- REST19002-900
- REST19003-900
- REST19005-271
- REST19011-853
- REST19005
- REST19006
- REST19007
- REST19008
- REST19023
- REST19001
- REST19002
- REST19040
- REST19003
- REST19004
- REST19019
- REST19020
- REST19021
- REST19022
- REST19000
- REST19013-900
- REST19014-900
- REST19017-291
- REST19019-810
- REST19016
- REST19017
- REST19018
- Temperaturlimit:
- 250 °C
- 250 °C
- 250 °C
- 250 °C
- 250 °C
- 250 °C
- 250 °C
- 300 °C
- 300 °C
- 300 °C
- 350 °C
- 250 °C
- 350 °C
- 350 °C
- 350 °C
- 250 °C
- 165 °C
- 300 °C
- 300 °C
- 0 - 175 °C
- 300 °C
- 350 °C
- 275 °C
- 165 °C
- 165 °C
- 165 °C
- 300 °C
- 250 °C
- 250 °C
- 275 °C
- 275 °C
- 275 °C
- 275 °C
- 275 °C