Bulk adsorbents, Florisil® PR
Catalog # REST26135
Supplier: Restek
CAS Number:
Specifications
- VE:1
- Bezeichnung:Florisil® PR bulk adsorbent, 60/100 mesh
- Environmentally Preferable:
Specifications
About this item
Florisil® PR is commonly used to remove polar interferences from pesticide residues. This bulk material is ideal for labs packing their own extraction cartridges for pesticide residue extractions.
- Pesticide residue grade
Packaging: Packaged in glass containers.