AuftragsverfolgungKontakt
Pure-Fit® SIB® Smooth Inner Bore Systems, Saint-Gobain Performance Plastics | Avantor
Pure-Fit® SIB® Smooth Inner Bore Systems, Saint-Gobain Performance Plastics
Artikelnr.
Pure-Fit® SIB® Smooth Inner Bore Systems, Saint-Gobain Performance Plastics
Artikelnr.
Anbieter:  
Pure-Fit® SIB® Smooth Inner Bore Systems, Saint-Gobain Performance Plastics
Pure-Fit® SIB® Smooth Inner Bore Systems, Saint-Gobain Performance Plastics
Product Details & Documents
Pure-Fit SIB is a smooth inner bore (SIB) hose-barb connector that provides a seamless transition between tubing and fittings throughout the fluid path.
  • Eliminates entrapment and leak points that occur with traditional fitting systems
  • Complete 360° compression along the entire fitting system
  • No excessive voids at the interface or in the fluid path
  • Utilizes the patented BarbLock retention and seal system
  • Compatible with Sani-Tech, C-Flex and PharMed flexible tubing
Available in both animal-free polypropylene (PP) material, which is gamma stable and polyvinylidene fluoride (PVDF) material, which is both autoclavable and gamma stable.
Typical areas of applications include biopharmaceutical manufacturing, media hydration, filtration, cell culture/harvest, product sampling and downstream activities.
 : Meets all USP Class VI and ISO criteria.