Anti-CD38 Mouse Monoclonal Antibody (APC-C750) [clone: HB7]
Catalog # ABNOMAB13894
Supplier: Abnova
Specifications
- Antikörper-Typ:Primär
- Antigen-Symbol:CD38
- Klonalität:Monoklonal
- Klon:HB7
- Konjugation:APC (Allophycocyanin)-C750
- Durchflusszytometrie:Yes
- Wirt:Mouse
- Isotyp:IgG1 kappa
- Reaktivität:Human
- Gen-ID:952
- Größe:100 reactions
- VE:100 Assays
Specifications
About this item
Mouse monoclonal antibody raised against human CD38.
Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HB7
Conjugation: APC-C750
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human