Anti-CD8A4 Mouse Monoclonal Antibody (PerCP) [clone: 143-44]
ABNOMAB13762
: Abnova
- Antikörper-Typ:Primär
- Antigen-Symbol:CD8A4
- Klonalität:Monoklonal
- Klon:143-44
- Konjugation:PerCP (Peridinin-Chlorophyll Protein Complex)
- Durchflusszytometrie:Yes
- Wirt:Mouse
- Isotyp:IgG1
- Reaktivität:Human
- Gen-ID:925
- Größe:100 reactions
- VE:100 Assays
Mouse monoclonal antibody raised against native human CD8A.
Type: Primary
Antigen: CD8A4
Clonality: Monoclonal
Clone: 143-44
Conjugation: PerCP
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human