Anti-CD7 Mouse Monoclonal Antibody (APC) [clone: HIT7]
ABNOMAB13756
: Abnova
- Antikörper-Typ:Primär
- Antigen-Symbol:CD7
- Klonalität:Monoklonal
- Klon:HIT7
- Konjugation:APC (Allophycocyanin)
- Durchflusszytometrie:Yes
- Wirt:Mouse
- Isotyp:IgG1
- Reaktivität:Human
- Gen-ID:924
- Größe:100 reactions
- VE:100 Assays
Mouse monoclonal antibody raised against human CD7.
Type: Primary
Antigen: CD7
Clonality: Monoclonal
Clone: HIT7
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human