Anti-CD6C10 Mouse Monoclonal Antibody (APC) [clone: MAE-1C10]
Catalog # ABNOMAB13749
Supplier: Abnova
Specifications
- Antikörper-Typ:Primär
- Antigen-Symbol:CD6C10
- Klonalität:Monoklonal
- Klon:MAE-1C10
- Konjugation:APC (Allophycocyanin)
- Durchflusszytometrie:Yes
- Wirt:Mouse
- Isotyp:IgG1
- Reaktivität:Human
- Gen-ID:923
- Größe:100 reactions
- VE:100 Assays
Specifications
About this item
Mouse monoclonal antibody raised against human CD6.
Type: Primary
Antigen: CD6C10
Clonality: Monoclonal
Clone: MAE-1C10
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human