Visual colour kits, alkalinity, AL-AP
Catalog # 705-0135
Supplier: Hach
CAS Number:
Specifications
- VE:1 KIT
- Bezeichnung:Alkalinity (CaCO₃)
- Parameter:Alcalinité (libre et totale en CaCO3)
- Tests pro Kit:100
- Messbereich:5 - 100 mg/l
Specifications
About this item
Caused by the presence of carbonates, bicarbonates, hydroxides, and other dissolved salts, alkalinity is important to food and beverage processors, boiler operators, and aquatic biologists.
- Kit utilises drop count titration method
- Simple, economical titration method
Delivery: Rugged kit contains all required reagents, solutions, and glassware.