Stativplatten, mit M10-Gewinde
Supplier: RSG
Jump to:
About this item
Retort stand bases made of stainless steel (Remanit 4301). With off-centre M10 thread.
2 Options Available Below
Product Details & Documents
Retort stand bases made of stainless steel (Remanit 4301). With off-centre M10 thread.
Product Family Options
Product Information