Test kits, sodium hypochlorite, CHECKIT®
Catalog # LOVI147490
Supplier: Lovibond
CAS Number:
Specifications
- VE:1 KIT
- Bezeichnung:Sodium hypochlorite test kit
- Messbereich:2 - 18%
Specifications
About this item
CHECKIT® Comparator is a compact and handy colorimetric unit which is suitable for both mobile and static analysis work.
- Easy operation and exact reagent dosing
- Tablet reagents have minimum guaranteed shelf life of 5/10 years
The operating instructions provide a step-by-step explanation of conducting the water test to achieve reliable and accurate measurements in the minimum of time.
Delivery information: Test kit includes CHECKIT® discs, cells, stirring rod and AQUALYTIC® tablet reagents (for 30 tests) for the desired test supplied in a sturdy plastic case.