Gas filters, charcoal
HEWL6810008600
: Agilent
- Bezeichnung:Argon gas filter, small, for 700 and vista series ICP-OES
- VE:1
Gas filters are specifically designed to remove impurity gases, moisture and hydrocarbons from the ICP-OES gas supplies.
- Deliver clean gases for accurate analyses
- Both inlet and outlet connectors are equipped with stainless steel frits to prevent particulates from entering the gas stream
- Easily replaced
This single combination filter removes water, trace impurity gases and organic compounds. No visual indicator. Installation date tag provided to provide a visual reminder when replacement required.