ContactUsLinkComponent
Ultra-low temperature freezers, upright models, −86 °C
Ultra-low temperature freezers, upright models, −86 °C
Supplier:  DAIREI EUROPE
Certificates
undefined
Ultra-low temperature freezers, upright models, −86 °C
Supplier:  DAIREI EUROPE
Specifications
  • Catalog No:
  • DAIRDAI1416
  • DAIRDAI1408
  • Modell:
  • ULUF 750
  • ULUF 450
  • Best.-Nr.:
  • DAIRDAI1416
  • DAIRDAI1408
  • Stromzufuhr:
  • 230 V, 50/60 Hz
  • 230 V, 50/60 Hz