NUCLEODUR® SiOH is an unmodified silica phase for HPLC.
- High purity spherical silica gel
- Very polar phase for NP chromatography
- Separation mechanism based on polar interactions
Recommended applications are separations of polar and midpolar compounds under normal phase conditions such as dyes from fuel oil (heptane extract).
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).
VP guard columns are used for preparative VarioPrep columns with the following guard column holders:
8,0×10 mm (Int.Ø×L) guard column for 8,0 and 10,0 mm (Int.Ø) preparative column with holder MANA718251.
16,0×10 mm (Int.Ø×L) guard column for 16,0 and 21,0 mm (Int.Ø) preparative column with holder MANA718256.
32,0×15 mm (Int.Ø×L) guard column for 32,0 and 40,0 mm (Int.Ø) preparative column with holder MANA718253.
50,0×15 mm (Int.Ø×L) guard column for ≥50,0 mm (Int.Ø) preparative column with holder MANA718255.
- Catalog No:
- MANA760012.46
- MANA762007.100
- MANA760007.46
- MANA762077.100
- MANA761967.30
- MANA761966.30
- MANA762078.100
- MANA762077.210
- MANA760007.40
- MANA760173.46
- MANA762094.80
- MANA760172.46
- MANA762094.160
- MANA762078.210
- MANA762007.210
- MANA760023.46
- pH-Bereich:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- USP-Code:
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3