Kugelfallprüfer
Impact test acc. to ASTM G14
Artikelnr. SIMESP1895
Anbieter: Industrial Physics
Springe zu:
Product Details & Documents
Dieser Kugelfallprüfer dient zur Bestimmung des Eindringwiderstands und der Flexibilität von Beschichtungen.
- Spirit level built-in for correct positioning
- Dual scale instrument equipped with a special guidance
- Distance between each impact is always according to the standards
Bestellinformation: Includes base plate, guide tube, release collar, punch (Ø = 16 mm) and V-notch vise with spring clamp to hold the pipe.
Dokumente
TQC Company Video
Videos
Spezifikationen
- Bezeichnung:Impact test acc. to ASTM G14
- Seq:03