Instrument dishes with lid, stainless steel
Artikelnr. KOEH2-1321
Lieferant: KOEHLER TECHNISCHE PRODUKTEN
Spezifikationen
- B×T×H:200×100×50 mm
- VE:1
Spezifikationen
Über diesen Artikel
Stainless steel dishes with radiused corners, dull-bright finish and dust-tight knob lids.
- Special corner shape for fast, easy cleaning
- Good chemical resistance
- Very good thermal resistance