Tyloxapol
Glasflasche, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Appearance: Clear to hazy yellow to amber Viscous liquid
- CAS Number:25301-02-4
- MDL Number:MFCD00149002
- Density:1,1 g/cm³
- Flash Pt:>113 °C
- Storage Temperature:Raumtemperatur
Documents
ACRO42237
SDS
Specifications
Specification Test Results
- Appearance:Clear to hazy yellow to amber Viscous liquid
- Infrared spectrum:Conforms
- pH:4.0 to 7.0 (5 % in water)
- Titration with NaOH:≥97.5 %
Specifications
- Packung:Glasflasche
- VE:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



