Baths and circulators, acrylic, SAHARA series
Lieferant: THERMO ELECTRON LED
Zertifikate
Über diesen Artikel
The SAHARA S3 series stainless steel heated circulators with acrylic baths are suitable for internal or external circulation.
- Feature bright display to view critical readings
- Thermostat can be turned and positioned on all four sides on the bath for easy viewing
It is rugged and corrosion-resistant for high temperature applications up to 150 °C.
Certifications: Units are RoHS-compliant (lead- and mercury-free) and UL/CSA compliant, assuring safe operation in the unlikely event of a fault condition.
Delivery: The instrument is supplied with communication cables, bridge with gasket and thumbscrews, fittings, clamps, pump plug for external circulation and 183 cm power cord.
Frequently Bought Together