Pierce™ Coomassie Plus (Bradford) Protein Assay Reagent
Artikelnr. PIER23238
Lieferant: Thermo Fisher Scientific
CAS-Nummer:
Spezifikationen
- VE:300 mL
- Bezeichnung:Coomassie plus (bradford) reagent, sufficient reagent to perform 200 standard assays or 1000 microplate assays
- Seq:01
Spezifikationen
Über diesen Artikel
The bradford protein assay is the preferred colorimetric assay for quantifying total protein concentration. The Pierce™ coomassie plus protein assay is a ready-to-use, reducing agent-compatible, improved bradford assay reagent to quickly measure (A595 nm) total protein concentration compared to a protein standard.
- Sensitive, fast and consistent protein assay kit
- Determine protein concentrations from 1 to 1500 μg/ml
- Ready-to-use dye binding reagent formulation
- Convenient microplate or cuvette format
- Compatible with reducing sugars, reducing substances and thiols
This modification of the well-known bradford method greatly reduces the tendency of coomassie reagents to give non linear response curves by a formulation that substantially improves linearity for a defined range of protein concentration. In addition, coomassie plus reagent results in significantly less protein-to-protein variation than is observed with other bradford-type coomassie formulations.