Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Zellkulturmedien
Dulbecco's modified eagle medium limiting media (DMEM-LM)
Dulbecco's modified eagle medium limiting media (DMEM-LM)
Dulbecco's modified eagle medium limiting media (DMEM-LM)
PIER30030
:
Thermo Fisher Scientific
:
Dulbecco's modified eagle medium limiting media (DMEM-LM)
PIER30030
:
Thermo Fisher Scientific
:
Medienformat:
Flüssigkeit
VE:
500 ml
Bezeichnung:
DMEM (-)L-leucine, (-)L-methionine with 4,5g/l glucose, 4,0 mM L-glutamine, sodium pyruvate and phenol red