Y-PER™ plus Yeast Protein Extraction Reagent, Dialysable
Catalog # PIER78999
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- VE:500 ml
- Bezeichnung:Y-PER-plus dialysable, yeast protein extraction reagent,
Specifications
About this item
This extraction reagent is formulated to effectively lyse yeast and bacterial cells to extract and solubilise proteins for analysis.
- No reducing or chelating agents
- Designed for room temperature storage
- Fast lysis completes after 20 minutes hands-off incubation
- Requires no rigorous mechanical disruption or lengthy enzymatic treatments
The reagent is used for Western blots, GST and histidine-tagged purification based applications. This fully dialysable formulation, allows complete exchange of the extracted protein into any buffer required by downstream techniques. The tris-based buffer has a very low ionic strength and does not include sodium chloride. The reagent is compatible with both the Thermo Scientific Pierce BCA and Coomassie Plus Protein Assays.