Molybdate reagent
Supplier: Hach
Certificates
About this item
Determination of high range phosphate (phosphorus) by the amino acid method.
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® pHenomenal®, pH-Elektroden
Supplier: VWR Collection
Cer(IV)sulfat Tetrahydrat, EMSURE® zur Analyse, Supelco®
Supplier: Merck
VWR®, Nitrilhandschuhe
Supplier: VWR Collection
VWR® Gesichtstücher
Supplier: VWR Collection
pH-Indikatorstäbchen, MQuant®, Supelco®
Supplier: Merck
Testpapiere pH, MQuant®, Supelco®
Supplier: Merck
Digitalbüretten, Titrette®
Supplier: Brand
Nitrat-Teststäbchen, MQuant®, Supelco®
Supplier: Merck
Peroxid-Teststäbchen, MQuant®, Supelco®
Supplier: Merck
Neßler-Reagenz
Supplier: Hach
Amino acid reagent
Supplier: Hach
VWR®, Universale pH-Elektroden
Supplier: VWR Collection
Laborkittel, Polyester/Baumwolle, 2852
Supplier: Portwest