ContactUsLinkComponent
Solution, cobalt chloride, for spectrophotometer
Solution, cobalt chloride, for spectrophotometer
  HACH1422249
 :  Hach
 :  
undefined
Solution, cobalt chloride, for spectrophotometer
  HACH1422249
 :  Hach
 :  
  • VE:
    500 mL
  • Bezeichnung:
    Cobalt chloride solution, 500 ml
Cobalt chloride solution for spectrophotometer linearisation.