VWR®, Disposable particle filtering respirators, N95
Artikelnr. VWRU89201-508
Lieferant: VWR Collection
Spezifikationen
- Typ:N-95
- Seq:01
- Größe:M/L
- VE:20
- Ausatemventil:-
Spezifikationen
Über diesen Artikel
The disposable lightweight N95 respirators combine excellent safety and comfort features to ensure a secure fit and a reliable protection.
- Protection against solid and liquid particles that do not contain oil
- Lightweight construction allows comfortable, extended use
- An adjustable noseclip provides a secure seal against the user's face
Certifications: NIOSH