VWR®, Sample/centrifuge tubes, with plug seal cap
Catalog # VWRU89012-778
Supplier: VWR Collection
Specifications
- Verpackt:50/bag, 10 bags/case
- Ausführung:Sterile
- Kapazität:30 ml
- Seq:01
- VE:500
Specifications
About this item
PP, clear, with translucent PP plug seal cap.
These tubes features moulded graduations and a leakproof plug seal cap designed to withstand 95 kPa of pressure. Skirted design enables tubes to stand upright, allowing for use without separate racks or stands.
Packaging: Bulk packaged with 500 separate caps.