Gas regulators
Chrome plated brass line gas regulator
Artikelnr. REST21666
Anbieter: Restek
Springe zu:
Product Details & Documents
Chromium plated brass line regulator.
- Oxidation resistant
- Same purity protection as high pressure cylinder regulators
Spezifikationen
- Bezeichnung:Chrome plated brass line gas regulator
- Cat. No.:REST21666
- Schnittstelle:1/4" FNTP
- Outlet pressure:0,00 - 3,45 bar