Order Entry
Austria
ContactUsLinkComponent
Anti-UBE2D1 Rabbit Polyclonal Antibody
Artikelnr. BOSSBS-8356R
Lieferant:  Bioss
Anti-UBE2D1 Rabbit Polyclonal Antibody
Artikelnr. BOSSBS-8356R
Lieferant:  Bioss

Spezifikationen

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Ubiquitin-conjugating enzyme E2D1
  • Antigen-Symbol:
    UBE2D1
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Gen-ID:
    7321
  • Antigen-Synonyme:
    Ubiquitin carrier protein|E2(17)KB 1|Ubiquitin-protein ligase|Ubiquitin-conjugating enzyme E2-17 kDa 1
  • Lagerungspuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Konzentration:
    1 ug/ul
  • Transporttemperatur:
    4 °C
  • Reinigung:
    Purified by Protein A.
  • Größe:
    100 μg
  • VE:
    100 µl

Spezifikationen

Über diesen Artikel

Accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. In vitro catalyzes 'Lys-48'-linked polyubiquitination. Mediates the selective degradation of short-lived and abnormal proteins. Functions in the E6/E6-AP-induced ubiquitination of p53/TP53. Mediates ubiquitination of PEX5 and auto-ubiquitination of CHIP, TRAF6 and TRIM63/MURF1. Ubiquitinates CHIP-associated HSP90AB1 in vitro. Lacks inherent specificity for any particular lysine residue of ubiquitin. Essential for viral activation of IRF3. Mediates polyubiquitination of CYP3A4.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: UBE2D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat