Order Entry
Austria
ContactUsLinkComponent
Anti-GGCX Rabbit Polyclonal Antibody
Anti-GGCX Rabbit Polyclonal Antibody
Catalog # BOSSBS-7759R
Supplier:  Bioss
Anti-GGCX Rabbit Polyclonal Antibody
Catalog # BOSSBS-7759R
Supplier:  Bioss

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Gamma-glutamyl Carboxylase
  • Antigen-Symbol:
    GGCX
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Gen-ID:
    2677
  • Antigen-Synonyme:
    VKCFD1
  • Lagerungspuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Konzentration:
    1 ug/ul
  • Transporttemperatur:
    4 °C
  • Reinigung:
    Purified by Protein A.
  • Größe:
    100 μg
  • VE:
    100 µl

Specifications

About this item

GGCX is a 758 amino acid multi-pass membrane protein. Localized to the membrane of the endoplasmic reticulum, GGCX functions to mediate the vitamin K-dependent carboxylation of glutamate residues on target proteins, thereby producing calcium binding gamma-carboxyglutamate (Gla) residues on these proteins and simultaneously converting vitamin K to vitamin K epoxide. GGCX exists as a monomer and, via its ability to modify glutamate residues, it accomplishes the post-translational changes that are necessary for the activity of all vitamin K-dependent proteins (such as blood coagulation and bone matrix proteins). Defects in the gene encoding GGCX are the cause of combined deficiency of vitamin K-dependent clotting factors 1 (VKCFD1) and PXE-like disorder with multiple coagulation factor deficiency, both of which are characterized by abnormal skin, blood or bone function.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: GGCX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat