ContactUsLinkComponent
Guard cartridge holder for HPLC columns
Artikelnr. WATEWAT046910
Lieferant:  WATERS
Guard cartridge holder for HPLC columns
Artikelnr. WATEWAT046910
Lieferant:  WATERS
Spezifikationen
  • Bezeichnung:
    Sentry guard cartridge holder kit, for 3,0, 3,9, and 4,6 mm ID columns
  • VE:
    1
Spezifikationen
Über diesen Artikel
The Sentry guard column holder is required for use with Sentry guard columns that have an I.D. of 3,0; 3,9 and 4,6 mm and a length of 20 mm.
This kit also contains a column joining tube assembly which allows your holder to connect with the analytical column. The sentry guard column holders can be used with any HPLC column regardless of the manufacturer.