Büchner-Trichter, mit Filterplatte
Catalog # SMIT8SFB/4/3
Supplier: SMITH SCIENTIFIC
Specifications
- Platte Ø:90 mm
- Porosität:3
- Seq:13
- VE:1
Specifications
About this item
Funnels, Büchner type, borosilicate glass 3.3, with integrated sintered glass filter plate.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes