Anti-TNFRSF11A Mouse Monoclonal Antibody [clone: 1E12]
Artikelnr. ABNOH00008792-M40
Lieferant: Abnova
Spezifikationen
- Antikörper-Typ:Primär
- Antigen-Name:tumor necrosis factor receptor superfamily, member 11a, NFKB activator
- Antigen-Symbol:TNFRSF11A
- Klonalität:Monoklonal
- Klon:1E12
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Mouse
- Isotyp:IgG1 kappa
- Reaktivität:Human
- Western blot:Yes
- Gen-ID:8792
- Antigen-Synonyme:ODFR|RANK|OFE|OSTS|FEO|LOH18CR1|OPTB7|CD265|PDB2|TRANCER
- Aminosäurezahl:69 to 152
- Sequence:IAPPLPCGPDEYLDSWNEEDKCLLHKVCDTGKALVAVVAGNSTTPRRCACTAGYHWSQDCECCRRNTECAPGLGAQHPLQLNKDTVCK
- Immunogen:TNFRSF11A (NP_003830.1, 69 a.a. ~ 152 a.a) partial recombinant protein with GST tag.
- Größe:100 μg
- VE:100 µG
Spezifikationen
Über diesen Artikel
Mouse monoclonal antibody raised against a partial recombinant TNFRSF11A.
Type: Primary
Antigen: TNFRSF11A
Clonality: Monoclonal
Clone: 1E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human