Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (PE (Phycoerythrin))
Catalog # ABNOMAB12225
Supplier: Abnova
Specifications
- Antikörper-Typ:Sekundär
- Antigen-Name:CD38 molecule
- Antigen-Symbol:CD38
- Klonalität:Monoklonal
- Klon:HIT2
- Konjugation:PE (Phycoerythrin)
- Durchflusszytometrie:Yes
- Wirt:Mouse
- Isotyp:IgG1 kappa
- Reaktivität:Human
- Gen-ID:952
- Antigen-Synonyme:ADPRC 1|ADPRC1
- Immunogen:Native purified CD38 from human thymocytes in foetus.
- Größe:100 Reactions
- VE:100 Assays
Specifications
About this item
Mouse monoclonal antibody raised against CD38.
Type: Secondary
Antigen: CD28
Clonality: Monoclonal
Clone: HIT2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human