Order Entry
Austria
ContactUsLinkComponent
 
Anti-ITGAM Mouse Monoclonal Antibody [clone: CD28.2] (FITC (Fluorescein Isothiocyanate))
  ABNOMAB12218
 :  Abnova
Anti-ITGAM Mouse Monoclonal Antibody [clone: CD28.2] (FITC (Fluorescein Isothiocyanate))
  ABNOMAB12218
 :  Abnova

 

  • Antikörper-Typ:
    Sekundär
  • Antigen-Name:
    integrin, alpha M (complement component 3 receptor 3 subunit)
  • Antigen-Symbol:
    ITGAM
  • Klonalität:
    Monoklonal
  • Klon:
    CD28.2
  • Konjugation:
    FITC (Fluorescein Isothiocyanate)
  • Durchflusszytometrie:
    Yes
  • Wirt:
    Mouse
  • Isotyp:
    IgG1 kappa
  • Reaktivität:
    Human
  • Gen-ID:
    3684
  • Antigen-Synonyme:
    CD11B|MAC-1|MO1A|MAC1A|SLEB6|CR3A
  • Immunogen:
    ITGAM
  • Größe:
    100 Reactions
  • VE:
    100 Assays

 

 

Mouse monoclonal antibody raised against ITGAX.

Type: Secondary
Antigen: ITGAM
Clonality: Monoclonal
Clone: CD28.2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human