Solid phase extraction disks, Resprep® Resin
Solid phase extraction disks, Resprep® Resin, 47 mm
Catalog # REST26023
Supplier: Restek
Jump to:
Product Details & Documents
Glass fiber disks embedded with styrene/DVB resin. The disks are used for chlorinated, benzidine-containing or nitrogen-containing pesticides.
Certifications: Meets the requirements of EPA methods 515.2 and 553.
Specifications
- Ø:47 mm
- Bezeichnung:Solid phase extraction disks, Resprep® Resin
- Seq:01