MSD source nut
Catalog # REST20643
Supplier: Restek
Specifications
- Bezeichnung:Detector MSD source nut
- Material:Messing
- VE:2
Specifications
About this item
This MSD source nut is made from brass. The 1,2 mm nut bore permits easy removal of ferrules with a standard tapered needle file. The material prevent thread-stripping on the transfer line, the design enhances ease of threading onto the transfer line and improves overall lifetime.