High-capacity split vent traps
Lieferant: Restek
Zertifikate
Über diesen Artikel
High-capacity split vent traps reduce the release of hazardous materials from the capillary split vent into the lab.
Delivery: Includes connecting lines and mounting kit.
Frequently Bought Together