Anti-ACC Rabbit Polyclonal Antibody
acetyl-coenzyme A is a component of the acetyl coenzyme A carboxylase complex. It is an enzyme that catalyzes carboxylation of acetyl-CoA to produce malonyl-CoA through its two catalytic activities, biotin carboxylase (BC) and carboxyltransferase (CT). ,acetyl-coenzyme A is a component of the acetyl coenzyme A carboxylase complex. It is an enzyme that catalyzes carboxylation of acetyl-CoA to produce malonyl-CoA through its two catalytic activities, biotin carboxylase (BC) and carboxyltransferase (CT).
Type: Primary
Antigen: ACC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Hyacinthus orientalis



