Anti-Cyt c Rabbit Polyclonal Antibody
AGRIAS08343A
: Agrisera
- Antikörper-Typ:Primär
- Antigen-Symbol:CYT C
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Reaktivität:T. weissflogii,C. reinhardtii,A. thaliana,N. tabacum,T. pseudonana
- Western blot:Yes
- UniProtKB:D7KMK0
- Immunogen:KLH-conjugated synthetic peptide derived from Arabidopsis thaliana cytochrome c protein sequence,
- Größe:50 µg
- VE:50 µG
Cytochrome c is located in inner mitochondrial membrane. It is a small heme protein which, unlike other cytochromes, is highly soluble. This protein is an essential component of the electron transport chain, where it undergoes oxidation and reduction without binding oxygen.,Cytochrome c is located in inner mitochondrial membrane. It is a small heme protein which, unlike other cytochromes, is highly soluble. This protein is an essential component of the electron transport chain, where it undergoes oxidation and reduction without binding oxygen.
Type: Primary
Antigen: CYT C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, C. reinhardtii, N. tabacum, T. weissflogii, T. pseudonana