ContactUsLinkComponent
PCR plates, 96-well, with half skirt, Multiply®
PCR plates, 96-well, with half skirt, Multiply®
Catalog # SARS72.1979
Supplier:  SARSTEDT
undefined
PCR plates, 96-well, with half skirt, Multiply®
Catalog # SARS72.1979
Supplier:  SARSTEDT
Specifications
  • Anz. Wells:
    96
  • Profil:
    Standard
  • Standring:
    Erhöhte Wells, Halbrand
  • Farbe der Wells:
    Klar
  • Barcodierung:
    Nein
  • VE:
    100