AuftragsverfolgungKontakt
Internal hand structure model | Avantor
Internal hand structure model
Internal hand structure model in three parts
Artikelnr. BINH1000349
Marketsource
Anbieter:   3B Scientific
Internal hand structure model
Artikelnr. BINH1000349
Anbieter:  3B Scientific
Marketsource
Internal hand structure model
Internal hand structure model
Internal hand structure model in three parts
Preis
992,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
This life size hand model shows the superficial and internal structures of the human hand including bones, muscles, tendons, ligaments, nerves, and arteries (superficial and deep palmar arches). The palmar aponeurosis and plate of the superficial tendons are removable from this high quality hand model.
Great for study of the anatomy of the human hand.
Spezifikationen
  • Bezeichnung:
    Internal hand structure model in three parts