Test bath
Test bath
SIMEVF8700
Accelerated corrosion test on panels and construction parts.
- Bezeichnung:Test bath
- Cat. No.:SIMEVF8700
- Genauigkeit:±0,1 °C
- B×T×H:302×240×150 (i.)
338×280×260 (o.) mm - Messbereich:5…100 °C
Accelerated corrosion test on panels and construction parts.