UV germicidal lamp
The UV germicidal lamp helps prevent contamination in research laboratories, food factories or in medical environment. These lamps emit 254 nm UV radiation, well known for their efficiency in the destruction of bacteria, moulds, yeasts and viruses.
- Exclusive UV Master® technology for high UV output
- Ondulex® reflector for optimum radiation
- Easy to handle thanks to a table support, handle and wall hook (optional)
Frequently Bought Together