Anti-MEF2C Rabbit Polyclonal Antibody
Artikelnr. ABNOPAB29248
Lieferant: Abnova
Spezifikationen
- Antikörper-Typ:Primär
- Antigen-Name:Myocyte enhancer factor 2C
- Antigen-Symbol:MEF2C
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunoChemie:Yes
- Western blot:Yes
- Kreuzadsorption:No
- Format:Liquid
- Gen-ID:4208
- Antigen-Synonyme:DEL5q14.3|C5DELq14.3
- Modification:phospho-Ser396
- Lagerungspuffer:PBS, pH 7,4 (without Mg(2+) and Ca(2+)), (50% glycerol, 0,02% sodium azide)
- Lagertemperatur:Store at –20 °C. Avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S396 of human MEF2C.
- Reinigung:Affinity purification
- Größe:100 μl
- VE:100 µl
Spezifikationen
Über diesen Artikel
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human MEF2C.
Recommended Dilutions: Immunohistochemistry: 1:50-1:100; Western Blot: 1:500-1000
Type: Primary
Antigen: MEF2C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: