Order Entry
Austria
ContactUsLinkComponent
Anti-FOS Mouse Monoclonal Antibody [clone: 2C9]
Artikelnr. ABNOH00002353-M63
Lieferant:  Abnova
Anti-FOS Mouse Monoclonal Antibody [clone: 2C9]
Artikelnr. ABNOH00002353-M63
Lieferant:  Abnova

Spezifikationen

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    FBJ Murine Osteosarcoma Viral Oncogene Homolog
  • Antigen-Symbol:
    FOS
  • Klonalität:
    Monoklonal
  • Klon:
    2C9
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Wirt:
    Mouse
  • Isotyp:
    IgG2a kappa
  • Reaktivität:
    Human
  • Western blot:
    Yes
  • Kreuzadsorption:
    No
  • Format:
    Liquid
  • Gen-ID:
    2353
  • Antigen-Synonyme:
    V-Fos FBJ Murine Osteosarcoma Viral Oncogene Homolog|AP-1|p55|C-FOS|G0/G1 Switch Regulatory Protein 7
  • Aminosäurezahl:
    1 to 100
  • Lagerungspuffer:
    1x PBS, pH 7,4
  • Sequence:
    MMFSGFNADYEASSSRCSSASPAGDSLSYYHSPADSFSSMGSPVNAQDFCTDLAVSSANFIPTVTAISTSPDLQWLVQPALVSSVAPSQTRAPHPFGVPA
  • Lagertemperatur:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    FOS (NP_005243.1, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
  • Größe:
    100 µg
  • VE:
    100 µg

Spezifikationen

Über diesen Artikel

Mouse monoclonal antibody raised against a partial recombinant FOS.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: FOS
Clonality: Monoclonal
Clone: 2C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human